PLOT columns, Rt®-Msieve 5A columns (fused silica)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19720 19721 19722 19723 19720-096 19722-6850 19723-106
REST19720EA 1035 EUR
REST19720 REST19721 REST19722 REST19723 REST19720-096 REST19722-6850 REST19723-106
PLOT columns, Rt®-Msieve 5A columns (fused silica)
Kolommen voor chromatografie
Molecular sieve 5A PLOT columns are designed for efficient separation of argon/oxygen and other permanent gases, including CH₄, C₂H₆, and CO.
  • Improve accuracy with sharp, symmetrical peaks for argon, oxygen, and carbon monoxide
  • Easily separate permanent gases at temperatures above ambient
  • PLOT technology reduces particle release, improving flow reproducibility and reducing downtime for maintenance
  • Stable to 300 °C
Special coating and deactivation procedures ensure chromatographic efficiency and the integrity of the porous layer coating. Molecular sieves have very high retention, allowing separations of permanent gases at temperatures above ambient. Additionally, the unique immobilisation process guarantees that the uniform particles remain adhered to the tubing - even after continuous valve-cycling.