Kwalitatief filterpapier, standaard klassen, klasse 595 en 595 ½, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10311610 10311611 10311612 10311687
512-3618EA 15.5 EUR
512-3618 512-3619 515-4505 515-5513
Kwalitatief filterpapier, standaard klassen, klasse 595 en 595 ½, Whatman™
Filters Filterpapier Kwalitatief filterpapier
A standard grade filter used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration of solids from digested environmental samples for iCP/AAs analysis).
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Grade 595: 4 - 7 µm
Very popular, thin filter paper, medium-fast with medium to fine particle retention. UsedMore Product Information
Order Now
Specificaties
Nr 595
Environmentally Preferable
Gewicht 68 g/m²
Dikte 150 µm
Partikelretentie 7 µm
Filtratiesnelheid 80 s (Herzberg)