pH electrodes for food applications
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
238080 238400 238418 238285
662-9208EA 305 EUR
662-9208 662-9219 662-0181 662-0176
pH electrodes for food applications
Elektroden pH-elektroden
Robust combination pH electrodes especially suited for use in media containing proteins or solid and semi-solid samples, such as meat and cheese.
  • TipTrode™ + FoodTrode™: Long-term, stable Everef system
  • Double Pore™: Two Single Pores make clogging of the liquid junction impossible
  • Electrodes with Polisolve electrolyte are maintenance-free
  • Electrodes with Protelyte electrolyte are refillable
pH: 0 to 14
Reference system: Everef (TipTrode™ + FoodTrode™); Ag/AgCl (Double Pore™)
Bestelinformatie: Supplied with serial number and certificate. ElectrodesMore Product Information