pH electrodes for general laboratory applications, Polilyte Lab
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
238403 242058 242060 242062 242059
662-9220EA 138 EUR
662-9220 662-9237 662-9236 662-9238 662-2080
pH electrodes for general laboratory applications, Polilyte Lab
Elektroden pH-elektroden
These robust combination pH electrodes are suitable for general laboratory applications, especially for measurements in emulsions and suspensions.
  • Single Pore junction prevents clogging of the liquid junction
  • Maintenance-free thanks to Polisolve electrolyte
  • Everef-B reference system for extended life time
pH: 0 to 14
Temperature: −10...+80 °C
Reference system: Everef-B
* Integrated temperature sensor NTC 30 kOhm
** Integrated temperature sensor Pt1000
Bestelinformatie: Supplied with serial number and certificate. Electrodes with S7 connector headMore Product Information