MINICYL regulators
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20610 20611
REST20610EA 266 EUR
REST20610 REST20611
MINICYL regulators
Gasregelaars
This compact general purpose regulator is constructed of lightweight aluminum with an elastomer diaphragm. It has many laboratory applications including air-drying glassware, sparging or evaporating solutions and controlling pneumatic valves.