Inserts for screw thread step vials
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21787 21788
REST21787EA 32 EUR
REST21787 REST21788
Inserts for screw thread step vials
flesjes
These inserts can be used with 4 ml WISP 48 screw thread step vials.