Inserts for crimp top and screw thread vials
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24517 24513 21776 21777 21779 21780 21782 24516 24518 24520 24692 24693
REST24517EA 230 EUR
REST24517 REST24513 REST21776 REST21777 REST21779 REST21780 REST21782 REST24516 REST24518 REST24520 REST24692 REST24693
Inserts for crimp top and screw thread vials
flesjes
These inserts are used with 2 ml, 11 mm crimp top and 2 ml, 9 mm short cap, screw thread vials.
Note: Big Mouth insert with glass flange is not to be used with 9 mm screw thread vials.