Screw thread vials and closures, short cap, 9 mm
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21140 21141 21142 21143 21154 21155
REST21140EA 38 EUR
REST21140 REST21141 REST21142 REST21143 REST21154 REST21155
Screw thread vials and closures, short cap, 9 mm
flesjes
Short cap screw thread vials fit all 2 ml, 12×32 mm, crimp-top vial-based autosamplers. Ideal for Agilent 7673, 7683, and 7693 autosamplers.