Bowls
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8017H-30 8017H-30S
SAMP8017H-30EA 81.6 EUR
SAMP8017H-30 SAMP8017H-30S
Bowls
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Mixing bowls, made of HDPE, ideal for laboratories, production and small scale work.
  • Robust construction
  • Individually wrapped
  • Food grade, FDA grade material of construction, EU 10/2011 compliant
  • BSE/TSE free