Internal tee, zero dead volume, Valco®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20154 20155
REST20154EA 123 EUR
REST20154 REST20155
Internal tee, zero dead volume, Valco®
Fittingen en connectoren
Internal tees are made of stainless steel are used to connect three lines. Suitable for 1,6 mm OD tubing.
Leveringsinformatie: Supplied with stainless steel ferrules.