Capillary column rinsing reservoir kit
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20612 20613
REST20612EA 265 EUR
REST20612 REST20613
Capillary column rinsing reservoir kit
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
This reservoir kit is used to restore the performance of bonded-phase capillary columns by dissolving and removing soluble, nonvolatile residue. The rinsing reservoir is equipped with 6,4 mm inlet and outlet connections and includes a built-in fritted disk to prevent particulate matter from contaminating the column.
Leveringsinformatie: Kit includes 50 ml reservoir, pressure regulator, fittings, ferrules, and tubing.