Metering gas valves, Parker Balston®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22201 22209
REST22201EA 0 EUR
REST22201 REST22209
Metering gas valves, Parker Balston®
Ventielen Luchtregelventielen
These metering gas valves reduce pressure between sample cylinder and GC injector.
Order Now
Specificaties
Environmentally Preferable