Mini-pots, with lid, stainless steel
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A426-100 A426-1000 A426-250 A426-500 A426-50 A426-25 A426-7
SAMPA426-100EA 194 EUR
SAMPA426-100 SAMPA426-1000 SAMPA426-250 SAMPA426-500 SAMPA426-50 SAMPA426-25 SAMPA426-7
Mini-pots, with lid, stainless steel
Reservoirs/Schalen/Potten Potten
Made of 316L stainless steel.
  • Crevice free interior
  • Screw on lid
  • Very good chemical and temperature resistance
Multi purpose small volume containers with satin finish.