Powder spatulas, disposable, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8095H-150S 8095H-150
SAMP8095H-150SEA 221 EUR
SAMP8095H-150S SAMP8095H-150
Powder spatulas, disposable, SteriWare®
Spatels
HDPE, white. These disposable spatulas feature a rounded handle and a robust blade.
  • Manufactured and packed in a controlled atmosphere (class 100000 cleanroom)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Approved for use in ATEX Zones 0, 1 & 2 (contact Sampling Systems for full details)
  • Disposable: No cleaning is required before or after sampling
Suitable for sterile sampling and dispensing quantities of powders in all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).