Bottles, narrow neck, with screw cap
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5.889.050 5.889.100 5.889.250
WITG5.889.050EA 1.1 EUR
WITG5.889.050 WITG5.889.100 WITG5.889.250
Bottles, narrow neck, with screw cap
Flessen Glazen flessen
Narrow neck bottles, made of soda glass, amber, with black screw cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.