Lepel spatel, brede vorm
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2013 2011 2010 2012
231-0125EA 225 EUR
231-0125 231-0123 231-0112 231-0124
Lepel spatel, brede vorm
Spatels
Stainless steel.
  • One end is flat and spatula-shaped, the other end is spoon-shaped
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
  • Non magnetic, bright finish