Extendable samplers, TeleSwab
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8011K-01 8011K-02
300-0420EA 4220 EUR
300-0420 300-0421
Extendable samplers, TeleSwab
Monsternemers Monsternemers voor vaste stoffen
The TeleSwab has an extendable handle, which allows to take swabs from high and difficult to reach places.
  • Environmental swabbing for use with large items of processing equipment
  • Extendable handle to the correct length for easy swabbing
  • Adapter allowing the swabs to be angled to the contours of the sampled item
Bestelinformatie: Swabs are not included with these kits.