Buffer, PBS (1X, Dulbecco's) powder pack
Leverancier: PanReac AppliChem
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A0964.9010
A0964.9050
A0964.9100
A0964.9010EA
35.5
EUR
A0964.9010
A0964.9050
A0964.9100
Buffer, PBS (1X, Dulbecco's) powder pack
Buffers
Buffers voor elektroforese
Running-buffers
Powder mixture to prepare PBS buffer (1X) according to Dulbecco.
Composed of di-sodium hydrogen phosphate anhydrous (1150 mg/l), potassium chloride (200 mg/l), potassium dihydrogen phosphate (200 mg/l) and sodium chloride (8000 mg/l).