Restriction enzymes, EcoRI, Fermentas
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ER0271 ER0272 ER0275
ER0271EA 49.4 EUR
ER0271 ER0272 ER0275
Restriction enzymes, EcoRI, Fermentas
Reagentia voor nucleïnezuren Restriction Enzymes
These EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37 °C in its own unique buffer. Exhibit 100% activity in the recommended buffer and reaction conditions.
  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient color-coded five buffer system and BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that sent in DNA preparations. Includes universal Tango buffer for double-digestions.