GC Columns, Rxi-PAH Columns (fused silica)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
49316 49317 49318
REST49316EA 1800 EUR
REST49316 REST49317 REST49318
GC Columns, Rxi-PAH Columns (fused silica)
Kolommen voor chromatografie
The Rxi-PAH GC columns were designed with a higher phenyl-content stationary phase that provides unique selectivity to separate important polycyclic aromatic hydrocarbons (PAH) for food safety that cannot be distinguished by mass spectrometry.
  • Ideal for EFSA PAH4 analysis—separates all priority compounds: benz[a]anthracene, chrysene, benzo[b]fluoranthene, and benzo[a]pyrene
  • Best resolution of chrysene from interfering PAHs, triphenylene and cyclopenta[cd]pyrene
  • Complete separation of benzo [b], [k], [ j], and [a] fluoranthenes
  • Thermal stability allows analysis of low-volatility dibenzo pyrenes