Grease, high vacuum, Dow Corning®
Leverancier: DOW CORNING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1597418_U DC970V
DOWC1597418_UEA 88.5 EUR
DOWC1597418_U DOWCDC970V
Grease, high vacuum, Dow Corning®
Smeermiddelen
This silicone-based clear translucent grey coloured compound is used as sealant and lubricant for vacuum systems.
  • Water-resistant
  • Bleed (200 °C, 24 hours) ˂0,5%
  • Evaporation (200 °C, 24 hours) ˂2 %
  • Has shelf life of 1800 days
Worked penetration: 230 mm/10
Unworked penetration: 175 mm/10 to 210 mm/10