Reverse frits
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4322-0100-144 4322-0100-294
NEUB4322-0100-144EA 411 EUR
NEUB4322-0100-144 NEUB4322-0100-294
Reverse frits
Filters Voorkolom filters en frits
Ideal for the filtration of substances sensitive to oxidation and hydrolysis. With filter plate, one-way cock and two standard ground-nuts.