Malaria HSP

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB81527-100 ORB81527-500
BIRBORB81527-100EA 0 EUR
BIRBORB81527-100 BIRBORB81527-500
Malaria HSP
Eiwitten en peptiden

Order Now

Specificaties

Protein/peptide name Malaria HSP
Environmentally Preferable
Species
Bron
Zuiverheid Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
Formulering 150mM Imidazole, pH 8.0, 150mM NaCl, 150mM Sodium Phosphate and 50% glycerol.
Storage conditions Five years frozen. One month in solution at room temperature.
Tested applications SDS-PAGE

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR