Kwantitatief filterpapier, asvrije klassen, klasse 589/2 en 589/2 ½, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10300102 10300103 10300106 10300107 10300108 10300109 10300110 10300111 10300112 10300114 10300120 10300143 10300145
515-3214EA 59.3 EUR
515-3214 515-3213 512-0608 515-3200 515-3201 512-3607 512-3608 512-3609 512-3610 512-3611 515-3207 512-0401 512-0609
Kwantitatief filterpapier, asvrije klassen, klasse 589/2 en 589/2 ½, Whatman™
Filters Filterpapier Kwantitatief filterpapier
Whatman Grade 589/2 filters from Cytiva are used in routine quantitative analyses, including analysis of aqueous suspension in paper, flour analysis, and sand content quantification.
  • High purity (<0,01% ash)
  • Retention of medium fine precipitates, such as grains of sand
  • Herzberg filtration speed of 140 s*
  • White ribbon filter
*The time taken to filter 100 ml water (20 °C) through a filter area of 10 cm² at a constant pressure of 5 cm water column.
Whatman quantitative filters from Cytiva's business are designed for gravimetric analysis. Grade 589/2 Whatman ashless filter paper can also be purchased as pre-pleated 589/2½ sheets.
Grade 589/2 ashless filter paper can be used when medium fine precipitates are being collected and papers must be ashed to support analysis. Grade 589/2 is suitable for a variety of routine quantitative analysis methods, such as determination of the sand content in foodstuffs, flour analysis, and aqueous suspension analysis in the paper industry.