Nitrogen generators, precision nitrogen trace
Leverancier: PEAK SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
62-0600 62-0250 62-1000
564-3001EA 8680 EUR
564-3001 564-3000 PEAK62-1000
Nitrogen generators, precision nitrogen trace
Gasgeneratoren Stikstofgeneratoren
These precision nitrogen trace generators produce "zero nitrogen" on demand from a compressed air source.
  • Suitable for carrier gas, make up gas and sample preparation
  • Regenerative CMS columns remove oxygen and moisture
  • Catalyst chamber to remove hydrocarbons (as methane) to <0,05 ppm with no expensive catalyst chamber replacements
Maximum pressure: 5,5 bar
Min/ max operating temperature: 5 to 35 °C
Gas outlet: 1×¹/₄" BSPP
Purity: >99,9995%
Weight: 21 kg (26 kg for the 1,00 l/min flow rate model)