VWR® Rota® Duo, pH indicator papers
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85403.600EA 63.4 EUR
85403.600 85404.600 85405.600 85406.600 85407.600 85408.600
VWR® Rota® Duo, pH indicator papers
Testpapieren pH-indicatorstrips
Duo indicator papers show two different colours for each pH value.
  • Easy to use for qualitatively accurate readings
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
  • Narrow range papers with graduation intervals of 0.3 pH units on the colour scale
  • Wide range papers with graduation intervals up to 1 pH unit on the colour scale
Leveringsinformatie: Supplied in plactic reels that ensure protection and long-term stability. Reel of 5 m length, width: 10 mm