Test strips, peracetic acid, Dosatest
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85437.601EA 39.6 EUR
85437.601 85436.601 85438.601
Test strips, peracetic acid, Dosatest
Milieutestkits
A ready to use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection process. Semi-quantitative results can be obtained between 10 and 120 seconds
Colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Verpakking: Container with 100 test strips 6×95 mm.