Mechanical lockout system, Pro-Lock II™
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
225204 225203 225205
BRDY225204EA 37.86 EUR
BRDY225204 BRDY225203 BRDY225205
Mechanical lockout system, Pro-Lock II™
Veiligheidssloten
This lockout is a complete isolation safety system, ideal for securing valves or industrial machinery.
  • Can only be opened and locked by using a special tool (not included)
  • Cable is made of stainless steel coated in PVC
Max. shackle diameter: 6,5 mm
Min. shackle clearance: 20,00 mm
Max. wire/cable diameter: 3,5 mm
Cable length: 1,5 m
Bestelinformatie: Supplied without operating tool which needs to be ordered separately.