Precision saccharometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3292108S 3292109S 3292110S 3292107S 3292111S 3292112S 3292113S
SCNL3292108SEA 130 EUR
SCNL3292108S SCNL3292109S SCNL3292110S SCNL3292107S SCNL3292111S SCNL3292112S SCNL3292113S
Precision saccharometers
Hydrometers
Saccharometers for beer and wort according to Plato with blue filling thermometer.
  • Suitable for calibration
Error limit: ±0,1 °Plato
Reference temperature: 20 °C
Bestelinformatie: Available with works calibration certificate or DAkkS calibration certificate on request.
Verpakking: In a square plastic sleeve.