PLOT columns, Rt-Q-BOND columns (fused silica)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19744 19742 19765 19741
REST19744EA 857 EUR
REST19744 REST19742 REST19765 REST19741
PLOT columns, Rt-Q-BOND columns (fused silica)
Kolommen voor chromatografie
Rt®-Q-BOND are nonpolar PLOT columns incorporating 100% divinylbenzene.
  • Analysis of C1 to C3 hydrocarbons as well as isomers and alkanes up to C12
  • Use for analysis of oxygenated compounds and solvents
  • Maximum temperature of 300 °C
High retention for CO₂ simplifies gas analysis. CO₂ and methane separated from O₂/N₂/CO (Note: O₂/N₂/CO not separated at ambient temperature)
Similar phase(s): HP PLOT Q, CP-PoraPLOT Q, CP-PoraBOND Q, Supel-Q-PLOT, AT-Q.