MicronQuick is based on the QuickStar micro cloth from Vileda Professional, developed to an even higher level of cleaning performance level. The Micron Fibre technology provides the finest split microfibres on the market, for deeper and more effective cleaning.
- Up to 99,99 % bacteria removal (proven by independent test institute (BMA))
- 25% improved cleaning performance (tested by the Fraunhofer Institute)
- Streak-free cleaning, no water residue left on the surface
- Lint free, improved wear resistance
- Easy to wring out and rinse
- Washable at 95 °C
MicronQuick has a very low friction action, which improves ergonomics. Due to the cloth's low volume, laundering costs are optimised and this combined with several hundred washing cycles means lower overall costs. MicronQuick is colour coded for maximum hygiene.