Pneumatic lockouts, Quick-Disconnect
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
064221 065645
BRDY064221EA 18.69 EUR
BRDY064221 BRDY065645
Pneumatic lockouts, Quick-Disconnect
Veiligheidssloten
These pneumatic quick-disconnect lockouts isolate pneumatic energy without the expense and inconvenience of installing in-line lockout valves.
  • Device is fitted to the male fitting, isolating equipment from all compressed air sources
  • Accommodates the majority of 6 mm, 9 mm and 12 mm male fittings
Max. shackle diameter: 7,84 mm
Min. shackle clearance: 20 mm