Single stage ultra high purity chrome plated brass gas regulators
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20646 20647 20648
REST20646EA 802 EUR
REST20646 REST20647 REST20648
Single stage ultra high purity chrome plated brass gas regulators
Gasregelaars
These chrome plated brass body ultra high purity gas regulators are ideal for sensitive GC applications using MS, PID and ECD detection methods. It features reduced internal dead volume relative to stainless steel bodies.
  • Oxidation-resistant chrome plating maintains a like-new appearance
  • Identical gas purity protection as with dual-stage gas regulators
  • Use when there is secondary pressure regulation downstream
  • Outlet assembly: 316 L stainless steel diaphragm valve and ¹/₄" tube fitting