Standard hazard warning signs
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
253254 827727 829570 829719 826771 830619 816674 828696 827651 827807 827649 827798 829727 830017 826762 827645 827647 829867 828831 826906 826910 255215 827355 826615 830458 827794 831372 828391 829274 829278 827667 827662 829868
BRDY253254EA 12.5 EUR
BRDY253254 BRDY827727 BRDY829570 BRDY829719 BRDY826771 BRDY830619 BRDY816674 BRDY828696 116-0942 BRDY827807 BRDY827649 116-0945 116-0943 BRDY830017 116-0947 BRDY827645 BRDY827647 BRDY829867 BRDY828831 BRDY826906 116-0944 116-0948 116-0949 116-0941 BRDY830458 BRDY827794 BRDY831372 BRDY828391 BRDY829274 116-0946 BRDY827667 BRDY827662 BRDY829868
Standard hazard warning signs
Pictogrammen
Warning signs must have a black pictogram on a yellow triangle with a black border. Hazard signs must be displayed to warn people to be careful or to take precaution where hazards exist.
  • Used for a wide range of applications
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Thickness: 1,5 mm
  • Non-adhesive