Capillary installation gauges
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22330 22335
REST22330EA 116 EUR
REST22330 REST22335
Capillary installation gauges
Uittrek- en invoeggereedschap
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
  • Prevents crushed column ends
  • Seats graphite ferrules onto column for consistent installations