Macrocellen met PTFE-stop voor absorptiemetingen
Leverancier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
110-10-46 110-5-46 110-5-40 110-20-40 110-40-40 110-1-46 110-1-40 110-10-40 110-2-40 110-10-K-40 110-50-40 110-5-K-46
634-9064EA 379 EUR
634-9064 634-9063 634-9053 634-9055 634-9056 634-9061 634-9051 634-9054 634-9052 634-0854 634-9057 HELL110-5-K-46
Macrocellen met PTFE-stop voor absorptiemetingen
Cuvetten Cuvetten voor spectrofotometers
Hellma® high precision quartz glass macro cells for absorption measurements in the UV/Vis/NIR range, 1 to 50 mm path length, with stopper.
  • Cells made of extended range quartz glass or high-performance quartz glass
  • Wavelength range from 200 to 3500 nm
  • Path length: 1 to 50 mm
  • With PTFE stopper
  • Volume: 350 to 17500 μl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism.