Replacement plunger assembly for autosampler syringes
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21218 21284
REST21218EA 112 EUR
REST21218 REST21284
Replacement plunger assembly for autosampler syringes
Spuitplunjer
This replacement plunger assembly with PTFE-tipped is ideal for Agilent 7673, 7683, 7693A, and 6850 autosampler syringes.