Gas-tight syringes, PTFE tip (SGE)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24725 24726 24727 24728 24729 24730 24734 24735 24737 24738 24739 24741 24742 24743 24750 24752 24755 24757 24759 24760 24761 24762
REST24725EA 0 EUR
REST24725 REST24726 REST24727 REST24728 REST24729 REST24730 REST24734 REST24735 REST24737 REST24738 REST24739 REST24741 REST24742 REST24743 REST24750 REST24752 REST24755 REST24757 REST24759 REST24760 REST24761 REST24762
Gas-tight syringes, PTFE tip (SGE)
Spuiten Injectiespuiten voor chromatografie
These gas-tight syringes with interchangeable barrels and plungers and tips extend performance and increase cost effectiveness.
  • Suitable for gases and liquids
  • High accuracy of dispensed volumes
Bestelinformatie: Needles have to be ordered separately for syringes without needles.