Injection needles
Leverancier: ROMETSCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13205-DZ 13205-07050
ROMT13205-DZEA 167 EUR
ROMT13205-DZ ROMT13205-07050
Injection needles
Injectienaalden
Rust-proof, with nickel-plated Luer lock fitting, reusable
  • Cannulas for puncturing and injecting
  • Blunted point tip