Containers for plant tissue culture, PLANTCON™, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
092672002 092672102
ICNA092672002EA 42.3 EUR
ICNA092672002 ICNA092672102
Containers for plant tissue culture, PLANTCON™, MP Biomedicals
Buisjes Cultuurbuisjes
PlantCon™ container is a presterilised, disposable, plastic growth containers. It consists of a cover and a bottom, and features a vent for gas exchange, and a drip control lip to recycle condensation.
  • Unbreakable container, safe to use
  • Presterilised, it is easy to use
  • Clear plastic provides excellent light transmission
  • Stackable
The cover can be snapped on tight, for long-term culture, or loosely fitted during acclimation to external conditions prior to transplantation. These containers are specifically designed for optimal plant cell culture applications.