Tubing for rotarus® peristaltic pumps, Tygon® F-4040-A
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95219016 95219015 95219013 95219017 95219503 95219725 95219717 95219710 95219703 95219522 95219517 95219510
HIRS95219016EA 189 EUR
HIRS95219016 HIRS95219015 HIRS95219013 HIRS95219017 HIRS95219503 HIRS95219725 HIRS95219717 HIRS95219710 HIRS95219703 HIRS95219522 HIRS95219517 HIRS95219510
Tubing for rotarus® peristaltic pumps, Tygon® F-4040-A
Slangen Transfer Tubing
These tubes are used for hydrocarbons and mineral oil products due to its low gas permeability, ozone resistant.
  • Temperature resistance from –40 to 75 °C
  • Hardness (Shore durometer A): 57