Tubing for rotarus® peristaltic pumps, Tygon® A-60-G
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95221037 95221038 95221036 95221035 95221019 95221018 95221017 95221016 95221015 95221013
HIRS95221037EA 305 EUR
HIRS95221037 HIRS95221038 HIRS95221036 HIRS95221035 HIRS95221019 HIRS95221018 HIRS95221017 HIRS95221016 HIRS95221015 HIRS95221013
Tubing for rotarus® peristaltic pumps, Tygon® A-60-G
Slangen Transfer Tubing
These tubes are used for industrial applications for its bending fatigue strength, good abrasion resistance and low gas permeability when compared to rubber tubes.
  • Temperature resistance from –60 to 135 °C
  • Hardness (Shore durometer A): 61