Cell culture media, powdered, Murashige and Skoog basal salt medium, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
092623020 092623022
ICNA092623020EA 47.6 EUR
ICNA092623020 ICNA092623022
Cell culture media, powdered, Murashige and Skoog basal salt medium, MP Biomedicals
Celcultuurmedia
Murashige and Skoog Basal Salt Medium is a widely used plant tissue culture growth medium. It contains macronutrients that include high levels of nitrate and organic additives such as agar, sugars, vitamins and growth regulators. It has proven effective in the culture of tissue derived from monocotyledons and dicotyledons.