IBC bunds, steel
Leverancier: CEMO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7392 7394 10613 10615 7393 7395 10614 10616
CEMG7392EA 1100 EUR
CEMG7392 CEMG7394 CEMG10613 CEMG10615 CEMG7393 CEMG7395 CEMG10614 CEMG10616
IBC bunds, steel
Absorptiematerialen en bescherming tegen morsen
IBC bunds are approved for storage of water-polluting and flammable substances.
  • Designed for the storage of large containers such as IBC, ASF- and ASP containers up to 1000 litres
  • Sumps are galvanised or painted, with removable grates
  • Equipped with galvanised, removable grates and 100 mm ground clearance for safe transport