Portable refractometers for clinical sports, PAL series
Leverancier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4442 4453 4454 4410
635-0246EA 308 EUR
635-0246 635-0257 635-0258 ATAG4410
Portable refractometers for clinical sports, PAL series
refractometers Draagbare refractometers
The PAL series with special scales are pocket instruments delivering rapid results.
Certificaten: IP 65 allows rinsing off the sample with running water.