Heater cartridge and platinum resistance thermometer (PRT) sensor for Agilent 5890 GCs
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22068 22069 23035
REST22068EA 424 EUR
REST22068 REST22069 REST23035
Heater cartridge and platinum resistance thermometer (PRT) sensor for Agilent 5890 GCs
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
This heater cartridge and PRT sensor can be used with 5890 FID and split/splitless weldments.