Canister filling stand
Leverancier: CEMO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8007 10100
CEMG8007EA 158 EUR
CEMG8007 CEMG10100
Canister filling stand
Staafstatieven
Filling stands, made from highly resistant stainless steel, suitable for acids and lyes.
  • Facilitates emptying of canisters up to 60 L
  • Cage secures mounting of canisters
  • Precise metering by swivelling of the canister