Capillary nuts
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22688 20375
REST22688EA 81 EUR
REST22688 REST20375
Capillary nuts
Fittingen en connectoren
These capillary nuts are ideal for Shimadzu 17A, 2010, and 2014 GCs.
The enhanced capillary nut design eliminates the slot, increasing lifetime and durability.