Environmental/HazMat cabinets, type 10/20 and 5/20
Leverancier: CEMO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8547 8552
CEMG8547EA 0 EUR
CEMG8547 CEMG8552
Environmental/HazMat cabinets, type 10/20 and 5/20
Meubilair Opslagkasten
Environmental/HazMat cabinet serves for safe storage of water and environmentally polluting substances within buildings.
  • Made of steel sheets and provided with folding doors
  • With general construction inspection approval no. Z-38.5-213
  • Lockable doors with 180° opening angle
  • Shelf heights can be adjusted in 15 mm steps
  • These cabinet support floors designed as a tray
The surface of the cabinet casing is sealed with a high quality powder coating (RAL 7035 light grey). The doors have ventilation slits top and bottom.
Certificaten: TÜV/GS tested.
Leveringsinformatie: Supplied with bottom and floor trays.