Injection port weldments
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22664 22665 22666 22667 22668 22669
REST22664EA 650 EUR
REST22664 REST22665 REST22666 REST22667 REST22668 REST22669
Injection port weldments
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Injection port weldments for Agilent GCs for use with purge and trap systems. This can easily attach your purge and trap with pre-installed low dead volume fittings.